Key Documents
BCS-535
CoolRack®
XT M24
About This Item
UNSPSC Code:
41103000
material
cylindrical wells
feature
for tubes 1.5 ‑ 2 mL
wells 24
Microfuge Tube Module
manufacturer/tradename
BioCision BCS-535
well diam. × depth
11.1 mm × 32.7 mm
General description
Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for cooling, (snap)freezing or heating/thawing samples.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)