Key Documents
BCS-529
CoolRack®
XT PCR96
About This Item
UNSPSC Code:
41103000
material
cone tip wells
feature
wells 96
PCR Plate, Strip Well, or Tube Module
manufacturer/tradename
BioCision BCS-529
well depth
13.2 mm
General description
Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for cooling, (snap)freezing or heating/thawing samples.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)