Key Documents
BCS-503G
CoolBox
2XT System, green
About This Item
UNSPSC Code:
41123304
feature
Includes CoolBox 2XT base, collar and lid, and two XT Cooling Cores (CoolRack or CoolSink sample modules purchased separately, accomodates 2)
manufacturer/tradename
BioCision BCS-503G
L × W × H
26.5 × 20 × 15 cm
color
green
General description
CoolBox 2XT System is a versatile ice-free alternative to ice buckets and gel-filled coolers. Cool, snap-freeze or transport samples without worry of contamination or temperature variation. Provides over 16 hours of cooling, without ice or power.
Magnetized collar secures tightly to the base providing a cold well for the sample module. Holds two thermo-conductive CoolRack or CoolSink sample modules, including some with automation-compatible features. Available in 4 colors - purple, green, orange and pink.
Applications include ice temperature cooling, maintaining frozen samples with optional XT Freezing Core (requires two), snap-freezing with dry ice in the base, short transport of temperature-sensitive samples, cooling or freezing in tissue culture hood, clean environment or other area where ice contamination is a concern
Legal Information
CoolBox is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)