Key Documents
BCS-502
CoolBox
XT System, purple
About This Item
UNSPSC Code:
41103000
feature
Includes CoolBox XT base, collar and lid, and one XT Cooling Core (CoolRack or CoolSink sample module purchased separately)
manufacturer/tradename
BioCision BCS-502
L × W × H
20 × 16 × 15 cm
color
purple
General description
CoolBox XT System is a versatile ice-free alternative to ice buckets and gel-filled coolers. Cool, snap-freeze or transport samples without worry of contamination or temperature variation. Includes CoolBox XT base, collar and lid, and one XT Cooling Core for 0.5 to 4°C cooling. Available in purple, green, orange or pink.
To use, pre-freeze the XT Cooling Core in a -20°C freezer until frozen. When ready to work, place frozen core into the base of the CoolBox XT and rest a CoolRack or CoolSink thermo-conductive tube or plate module on the core. Provides over 16 hours of 0.5 to 4°C cooling with lid closed and over 10 hours with the lid open. Magnetized collar secures tightly to the base providing a cold well for the sample module. Holds one thermo-conductive CoolRack or CoolSink sample module, including some with automation-compatible features.
Applications include ice temperature cooling, maintaining frozen samples with optional XT Freezing Core (requires one), snap-freezing with dry ice in the base, short transport of temperature-sensitive samples, cooling or freezing in tissue culture hood, clean environment or other area where ice contamination is a concern.
Legal Information
CoolBox is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)