Key Documents
BCS-265
CoolRack®
SV10
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
material
cylindrical wells
feature
for tubes 10 mL
wells 12
Cell Therapy Ampule Module
manufacturer/tradename
BioCision BCS-265
well diam. × depth
23.6 mm × 27.9 mm
General description
Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for cooling, (snap)freezing or heating/thawing samples.
Features and Benefits
Ideal for keeping sterile samples cool during cell therapy workflow.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)