Key Documents
BCS-2510
TruCool® Cryogenic Vials
capacity 1 mL, self-standing, sterile, internal-thread vials with auto-cap, case of 500 ea
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
sterility
sterile
feature
self-standing
packaging
case of 5 × 100 packs
case of 500 ea
manufacturer/tradename
BioCision BCS-2510
capacity
1 mL
tube diam. × H
12.5 mm × 43.0 mm
General description
TruCool leak-proof cryogenic vials feature a screw cap with a thermally-fused gasket to ensure a tight seal. The gasket is a co-molded thermoplastic elastomer (TPE) layer, which prevents leaking, slipping and risk of contamination. The gasket is 95kPa certified to provide a leak-proof seal. Cap accomodates auto-decapping equipment.
Cryogenic vials are sterile and individually barcoded with a unique identifier which can be read with common barcode readers. Ideal for use in CoolRack CF cryogenic vial modules and CoolCell alcohol-free cell freezing containers. Recommended for use in vapor phase LN2
Legal Information
TruCool is a registered trademark of BioCision LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)