BCS-2502
TruCool® Cryogenic Vials
capacity 2 mL, self-standing, sterile, external-thread vials with auto-cap, case of 500 ea
All Photos(1)
About This Item
UNSPSC Code:
41121704
NACRES:
NB.22
Recommended Products
sterility
sterile
feature
self-standing
packaging
case of 5 × 100 packs
case of 500 ea
manufacturer/tradename
BioCision BCS-2502
capacity
2 mL
diam. × H
12.5 mm × 47.5 mm
General description
TruCool leak-proof cryogenic vials feature a screw cap with a thermally-fused gasket to ensure a tight seal. The gasket is a co-molded thermoplastic elastomer (TPE) layer, which prevents leaking, slipping and risk of contamination. The gasket is 95kPa certified to provide a leak-proof seal. Cap accomodates auto-decapping equipment.
Cryogenic vials are sterile and individually barcoded with a unique identifier which can be read with common barcode readers. Ideal for use in CoolRack CF cryogenic vial modules and CoolCell alcohol-free cell freezing containers. Recommended for use in vapor phase LN2
Cryogenic vials are sterile and individually barcoded with a unique identifier which can be read with common barcode readers. Ideal for use in CoolRack CF cryogenic vial modules and CoolCell alcohol-free cell freezing containers. Recommended for use in vapor phase LN2
Legal Information
TruCool is a registered trademark of BioCision LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)