Key Documents
BCS-207
TruCool® Hinged CryoBox
81-Place, white, pack of 50 ea
About This Item
UNSPSC Code:
41123304
NACRES:
NB.22
feature
81-Place
packaging
pack of 50 ea
manufacturer/tradename
BioCision BCS-207
L × W × H
13.3 cm × 13.3 cm × 5.1 cm
color
white
General description
Standardize your cryogenic sample storage
  • Patented hinged lid ensures markings and samples remain in synch
  • Lid is easy to open when frozen and stays attached to base, minimizing contamination risk
  • Plastic 81-place grid is durable and adjustable, enabling the storage of wide diameter tubes
  • Laminated fiberboard box is water-resistant, durable and recommended for refrigerators, freezers, ultra-low freezer storage or liquid nitrogen vapor
  • Store up to 81 cryogenic vials (1 mL or 2 mL) or microtubes
  • Holds up to repeated freezing and thawing better than conventional cardboard boxes
  • Six bright colors allow sample segregation and are quickly visible in frosty freezers and storage tanks.
Legal Information
TruCool is a registered trademark of BioCision LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)