Key Documents
BCS-205
CoolRack®
Sleeves, pkg of 4 sleeves
About This Item
UNSPSC Code:
41122806
NACRES:
NB.22
packaging
pkg of 4 sleeves
manufacturer/tradename
BioCision BCS-205
color
assorted colors
General description
Colorful elastic sleeves brighten your lab and allow you to color-code your experiments, while providing a comfortable grip when handling cold , warm or wet CoolRack tube modules. Compatible with ice, dry ice and water baths.
Sleeves fit the following modules:
All 15-well CoolRack modules
All 30-well CoolRack modules
CoolRack 15ml
CoolRack 50ml
CoolRack VS13
CoolRack V13
CoolRack V16
Four colors: purple, green, orange, pink
Pack of 4, one of each color.
Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for cooling, (snap)freezing or heating/thawing samples.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)