Key Documents
BCS-190
BioCision TruCool® Tote Bag
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
feature
Insulated and water-proof sample carrying tote
manufacturer/tradename
BioCision BCS-190
L × W × H
18.5 × 6.5 × 11 in.
General description
This insulated tote is perfect for carrying temperature-sensitive samples between labs or clinical sites. Interior is insulated and waterproof making it suitable for use with ice or dry ice, along with CoolRack or CoolSink sample holders. Keep samples uniformly cold or frozen during short transit.
Legal Information
TruCool is a registered trademark of BioCision LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)