Key Documents
BCS-126
CoolRack®
CF15
About This Item
UNSPSC Code:
41103000
material
cylindrical wells
feature
wells 15
Cryogenic Vial Module
manufacturer/tradename
BioCision BCS-126
well diam. × depth
12.7 mm × 32.7 mm
General description
Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for cooling, (snap)freezing or heating/thawing samples.
Features and Benefits
"Gripping" wells for one-hand vial opening/closing.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)