Key Documents
BAH445770001
Scienceware® slide box
for 100 microscope slides, Slip-on lid
About This Item
UNSPSC Code:
41123302
material
plastic
feature
Holds 100 x slides 76x26 mm
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H445770001
General description
Supreme defense for standard 25 x 75mm (1" x 3") microscope slides
  • Heavy-walled, moisture- and insect-proof.
  • Slim, durable design
  • Easily identify slides with included numbered index on base and cover
  • Box walls are rugged and won′t bend, split, or chip.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)