Key Documents
BAF441820000
Slide Staining and Storage System
Staining Dish, polyoxymethylene
About This Item
UNSPSC Code:
12352200
material
polyoxymethylene
feature
1 Slide Rack
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F44182-0000
L × W × H
9.4 x 8.1 x 4.9 cm (3¹¹⁄₁₆ in.) × 8.1 cm (3³⁄₁₆ in.) × 4.9 cm (2 in.)
General description
Versatile slide rack and vessel for radiography gel staining and staining protocols. PMPA plastic construction makes the unit virtually shatter-proof, lightweight, with exceptional solvent and heat resistance for extended use in cytology and histology labs. The rack and dish can simply be cleaned and are highly resistant to standard stains used in staining protocols.
Conveniently stain and store gels
  • Polyoxymethylene-poly-acetal (PMPA) plastic construction
  • Excellent solvent and heat resistance
Choose from one of the most recent versions:
Certificates of Analysis (COA)