Key Documents
BAF441800000
Slide Staining and Storage System
Slide Rack, Holds 25 x slides 76x26 mm
About This Item
UNSPSC Code:
12352200
material
polyoxymethylene
feature
Holds 25 x slides 76x26 mm
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F44180-0000
size
9.2 cm (3.6 in.) × 7.8 cm (3 in.) × 2.0 cm (0.78 in.)
General description
Versatile slide rack and vessel for radiography gel staining and staining protocols. PMPA plastic construction makes the unit virtually shatter-proof, lightweight, with exceptional solvent and heat resistance for extended use in cytology and histology labs. The rack and dish can simply be cleaned and are highly resistant to standard stains used in staining protocols.
Conveniently stain and store gels
  • Polyoxymethylene-poly-acetal (PMPA) plastic construction
  • Excellent solvent and heat resistance
Choose from one of the most recent versions:
Certificates of Analysis (COA)