Key Documents
BAF409130000
Bel-Art® Inoculating Turntable
for 60 to 100 mm Petri Dishes
About This Item
UNSPSC Code:
12352200
material
polypropylene
diam. × H
5 in. × 2 in.
General description
Manually moved turntable creates virtually concentric circles of uniformly spread out bacterial colonies. This turntable can hold various sizes of Petri dishes -- 60, 90, 100mm. Ball bearings provide a steady, even, and prolonged spin time resulting in a rapid inoculation step and extended operating life.
Equally allocate bacteria across Petri Dishes with ease.
  • Ball bearings assure simple and graceful spinning
  • Lightweight, non-skid, durable and portable
  • Polypropylene construction
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)