BAF378511000
CUBE 2UBE® Tubes
polypropylene, pack of 1000 tubes
All Photos(2)
About This Item
UNSPSC Code:
41121800
material
colorless tube
polypropylene
polypropylene tube
round bottom
sterility
non-sterile
feature
graduations: no
packaging
pack of 1000 tubes
manufacturer/tradename
Bel-Art F37851-1000
capacity
2.2 mL
binding type
non-treated surface
General description
CUBE 2UBE Tubes offer rapid, convenient sample storage and media preparation with 2.2ml per tube in a 8 x 12, 96 well format. The patented CUBE 2UBE Tubes is compatible with most automated liquid handling systems and can be centrifuged up to 14,000 RPM. The sturdy polypropylene CUBE 2UBE Tube can be frozen to -40 deg C. CUBE 2UBE Tubes are compatible with many Robotic workstations. Boxed in rigid polypropylene. Each tube has a capacity of 2.2MLS or 211MLS per rack of 96 tubes.
Rapid, convenient sample storage and media preparation in 2.2ml wells
Rapid, convenient sample storage and media preparation in 2.2ml wells
- Standard 8 x 12, 96 well format
- Rugged polypropylene construction
Legal Information
CUBE 2UBE is a registered trademark of SP Scienceware, Division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)