BAF249890000
Bel-Art® Disposable Charcoal Cartridge
pack of 3 cartridges
About This Item
Recommended Products
packaging
pack of 3 cartridges
manufacturer/tradename
Bel-Art F24989-0000
General description
Safely absorb volatile 35S radioactive components during labeling experiments
- Ideal for use in incubators
- Disposabl
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service