Key Documents
BAF210120000
Bel-Art® Safety Cutter
About This Item
UNSPSC Code:
41122406
NACRES:
NB.35
packaging
pkg of 3 ea
manufacturer/tradename
Bel-Art F21012-0000
L × W × H
6 cm × 3 cm × 0.5 cm
General description
Easily and safely cut LabMat Bench Liner as well as plastic “clamshell” packaging, plastic bags, shrink-wrap, paper, and much more. Faster than scissors, safer than utility knives.
Features and Benefits
Blade will not cut fingers and does not need to be covered for safety
Small, ceramic blade will never rust and will stay sharp much longer than steel blades
Easily cut small areas of LabMat containing hazardous spills to minimize disposal costs associated with hazardous materials
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Labmat is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)