Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B9803

Sigma-Aldrich

Bone Morphogenetic Protein 5 human

recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >95% (SDS-PAGE)

Synonym(s):

BMP-5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in NSO cells

assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

subunit calculated mol wt ~18 kDa

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BMP5(653)

Biochem/physiol Actions

Cellular responses to BMP-5 is mediated by the formation of hetero-oligomeric complexes of type I and type II serine/threonine kinase receptors.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile, 0.1% TFA, with 5% trehalose and 50 μg of BSA per 1 μg of cytokine

Analysis Note

The biological activity is measured by its ability to induce alkaline phosphatase production in ATDC5 chondrogenic cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ebendal et al.
Journal of neuroscience research, 51(2), 139-146 (1998-02-20)
Transforming growth factors-beta (TGF-betas), activins, and bone morphogenetic proteins (BMPs) comprise an evolutionarily well-conserved group of proteins controlling a number of cell differentiation, cell growth, and morphogentic processes during development. The superfamily of TGFbeta-related genes include over 25 members in
A H Reddi
Nature biotechnology, 16(3), 247-252 (1998-04-07)
Morphogenesis is the developmental cascade of pattern formation and body plan establishment, culminating in the adult form. It has formed the basis for the emerging discipline of tissue engineering, which uses principles of molecular developmental biology and morphogenesis gleaned through
M Kawabata et al.
Cytokine & growth factor reviews, 9(1), 49-61 (1998-08-28)
Bone morphogenetic proteins (BMPs) are multifunctional cytokines, which are members of the transforming growth factor-beta (TGF-beta) superfamily. Activities of BMPs are extracellularly regulated by BMP-binding proteins, Noggin and Chordin. BMPs bind to two different types of serine-threonine kinase receptors, type
Elina V García et al.
Theriogenology, 84(4), 589-599 (2015-05-28)
Previous studies have reported that bone morphogenetic protein 5 (BMP5) is differentially expressed in the isthmus of bovine oviducts and it is present in the oviductal fluid. However, the specific action of this factor is unknown. To evaluate whether BMP5
Zhu Xishan et al.
Journal of experimental & clinical cancer research : CR, 30, 47-47 (2011-05-04)
Overwhelming evidence from leukemia research has shown that the clonal population of neoplastic cells exhibits marked heterogeneity with respect to proliferation and differentiation. There are rare stem cells within the leukemic population that possess extensive proliferation and self-renewal capacity not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service