Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B9554

Sigma-Aldrich

BDS-I

≥95%, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Product B9554 is not currently sold in your country. Contact Technical Service

Quality Level

assay

≥95%

form

lyophilized powder

mol wt

4715 Da

storage temp.

−20°C

Biochem/physiol Actions

Specifically blocks the rapidly inactivating Kv3.4 channel.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Major toxin in Anemonia sulcata

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Diochot et al.
The Journal of biological chemistry, 273(12), 6744-6749 (1998-04-18)
Sea anemone venom is known to contain toxins that are active on voltage-sensitive Na+ channels, as well as on delayed rectifier K+ channels belonging to the Kv1 family. This report describes the properties of a new set of peptides from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service