Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B8648

Magenta B-cap

for use in plant tissue culture, autoclavable

Synonym(s):

Container Cap, Magenta Cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

feature

autoclavable

application(s)

agriculture

General description

Designed for use in plant tissue culture laboratories using baby food culture jars (Product Codes V8630 and V0633). Unique design facilitates excellent light penetration and easy usage; autoclavable.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ethan T Pierce et al.
Planta, 249(5), 1551-1563 (2019-02-08)
A novel method for culturing ovules of Gossypium barbadense allowed in vitro comparisons with Gossypium hirsutum and revealed variable roles of microtubules in controlling cotton fiber cell expansion. Cotton fibers undergo extensive elongation and secondary wall thickening as they develop

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service