Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B8561

Sigma-Aldrich

Anti-BRCAA1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ARID4B, Anti-AT rich interactive domain 4B (RBP1-like), Anti-BCAA, Anti-RBBP1L1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~136 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARID4B(51742)
mouse ... Arid4b(94246)
rat ... Arid4b(84481)

Immunogen

synthetic peptide corresponding to residues 1046-1055 of human BRCAA1. This sequence is identical in mouse and rat

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service