B8554
Anti-Bax antibody, Mouse monoclonal
clone 2D2, purified from hybridoma cell culture
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2D2, monoclonal
form
buffered aqueous solution
mol wt
antigen 21 kDa (and possibly a dimer of 42 kDa)
species reactivity
human, bovine
should not react with
rat, mouse
concentration
~2 mg/mL
technique(s)
microarray: suitable
western blot: 1-5 μg/mL using cultured human breast adenocarcinoma MCF-7 cells, activated with dexamethasone
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... BAX(581)
Looking for similar products? Visit Product Comparison Guide
General description
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service