Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B8062

Sigma-Aldrich

Anti-Par6β antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PAR6B, Anti-PARD6B, Anti-Partitioning-defective protein 6, Homolog beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2-5 μg using MDCK cell lysate
indirect immunofluorescence: 2-5 μg/mL using paraformaldehyde-fixed MDCK cells
western blot: 0.5-1 μg/mL using lysates of MDCK cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PARD6B(84612)

Immunogen

synthetic peptide corresponding to amino acids 275-289 of human Par6β conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Dong et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 245(6), 653-666 (2016-03-19)
TMEM47 is the vertebrate orthologue of C. elegans VAB-9, a tetraspan adherens junction protein in the PMP22/EMP/Claudin family of proteins. VAB-9 regulates cell morphology and adhesion in C. elegans and TMEM47 is expressed during kidney development and regulates the activity
Qiao-Song Zheng et al.
Reproduction (Cambridge, England), 147(1), 45-52 (2013-10-17)
Spermatogenesis is a complex process involving the regulation of multiple cell types. As the only somatic cell type in the seminiferous tubules, Sertoli cells are essential for spermatogenesis throughout the spermatogenic cycle. The Wilms tumor gene, Wt1, is specifically expressed
Min-Kyung Yeo et al.
Translational oncology, 11(1), 109-115 (2017-12-09)
Partitioning defective (Par) proteins regulate cell polarity and differentiation. Par3, Par6β, and protein kinase Cζ (PKCζ), which are PAR complex members, have been shown to be associated with oncogenesis and progression. Herein, we report the expression pattern and clinical relevance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service