Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B7938

Sigma-Aldrich

BTA-EG4 hydrate

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N2O4S · xH2O
CAS Number:
Molecular Weight:
416.53 (anhydrous basis)
UNSPSC Code:
12352202

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white to brown

solubility

DMSO: >15 mg/mL

storage temp.

2-8°C

Biochem/physiol Actions

BTA-EG4 is a probe for β-amyloid aggregates and an inhibitor of protein-amyloid interaction.
BTA-EG4 is a probe for β-amyloid aggregates and an inhibitor of protein-amyloid interaction. BTA-EG4 inhibits several different Ab -binding proteins from associating with aggregated Ab peptides. It inhibits catalase-Aβ binding interactions in solution by forming protein-resistant surface coatings on aggregated Aβ (1–42) peptides. These coatings reduce the toxicity of aggregated Aβ, causing an increase in cell viability.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service