B7911
Monoclonal Anti-Phosphoserine−Biotin antibody produced in mouse
clone PSR-45, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Monoclonal Anti-Phosphoserine, Phospho Ser, Phospho serine, Phospho−Ser, Phospho−serine
About This Item
Recommended Products
biological source
mouse
conjugate
biotin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
PSR-45, monoclonal
form
buffered aqueous solution
technique(s)
direct ELISA: 1:50,000-1:100,000 using phosphoserine-BSA at 10 μg/ml, and ExtrAvidin-HRP at 2 μg/ml
dot blot: 1:200,000 using 40 ng phosphoserine-BSA/dot and ExtrAvidin-HRP at 1 μg/ml
isotype
IgG1
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Looking for similar products? Visit Product Comparison Guide
General description
Monoclonal Anti-Phosphoserine may be used for the identification of proteins containing phosphorylated serine both as the free amino acid or when conjugated to carriers such as BSA or KLH. The purified mouse immunoglobulin from ascites fluid is conjugated to biotinamidohexanoic acid N-hydroxysuccinimide ester. This covalent coupling of biotin to the immunoglobulin allows for the binding of avidin or streptavidin bearing a variety of different labels.
Specificity
Immunogen
application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service