Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B7032

Sigma-Aldrich

Boc-5-Cyclohexylstatine

≥98% (TLC)

Synonym(s):

Boc-(3S,4S)-4-amino-5-cyclohexyl-3-hydroxypentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H29NO5
CAS Number:
Molecular Weight:
315.41
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Product Name

Boc-5-Cyclohexylstatine, ≥98% (TLC)

assay

≥98% (TLC)

form

solid

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)NC(CC1CCCCC1)C(O)CC(O)=O

InChI

1S/C16H29NO5/c1-16(2,3)22-15(21)17-12(13(18)10-14(19)20)9-11-7-5-4-6-8-11/h11-13,18H,4-10H2,1-3H3,(H,17,21)(H,19,20)

InChI key

IJMMECIIPUTJIH-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Boger et al.
Journal of medicinal chemistry, 28(12), 1779-1790 (1985-12-01)
Analogues of the renin octapeptide substrate were synthesized in which replacement of the scissile dipeptide with (3S,4S)-4-amino-3-hydroxy-6-methylheptanoic acid (statine, Sta) transformed the substrate sequence into potent, transition-state analogue, competitive inhibitors of renin. Synthesis and incorporation of the cyclohexylalanyl analogue of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service