Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B6928

Sigma-Aldrich

Mouse Brain Extract

2 mg/mL in SDS-PAGE loading buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

form

solution

Quality Level

concentration

2 mg/mL in SDS-PAGE loading buffer

storage temp.

−20°C

Application

Mouse Brain Extract is Ready-To-use as a positive control in immunoblotting techniques (optimal concentration must be determined by user). Useful as a control for Anti-PKA RII, Anti-bNOS and other antibodies.

Other Notes

Whole tissue extract of mouse brain.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service