Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B6562

Sigma-Aldrich

Anti-Bif-1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Bax-interacting factor 1, Anti-Endophilin B1, Anti-SH3-containing protein SH3GLB1, Anti-SH3-domain GRB2-like endophilin B1, Anti-SH3GLB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

rat, mouse, human

concentration

~1.0 mg/mL

technique(s)

western blot: 2-4 μg/mL using using whole extracts of HEK-293T cells expressing human Bif-1, rat NRK, and mouse 3T3 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service