B6274
5-Bromo-4-chloro-3-indolyl phosphate dipotassium salt
phosphatase substrate
Synonym(s):
BCIP®, X-phosphate dipotassium salt
About This Item
Recommended Products
assay
≥99% (HPLC)
form
powder
solubility
water: 50 mg/mL, clear to hazy
storage temp.
−20°C
SMILES string
[K].OP(O)(=O)Oc1c[nH]c2ccc(Br)c(Cl)c12
InChI
1S/C8H6BrClNO4P.K.H/c9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14;;/h1-3,11H,(H2,12,13,14);;
InChI key
HPKPHSVUJYUSDD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Substrates
Legal Information
comparable product
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service