Butyl p-aminobenzoate-agarose is an agarose conjugate in buffered aqueous suspension used in affinity chromatography, protein chromatography and hydrophobic interaction methodologies. Butyl p-aminobenzoate-agarose has been used in studies to assess local anesthetic inhibition and activity of human serum albumin.
Physical form
Suspension in 0.05 M NaHCO3, 0.5 M NaCl, pH 8.0, containing preservative
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
The Journal of pharmacology and experimental therapeutics, 314(3), 1177-1186 (2005-06-01)
n-Butyl-p-aminobenzoate (BAB; butamben) is a long-duration anesthetic used for the treatment of chronic pain. Epidural administration of BAB is thought to reduce the electrical excitability of dorsal root nociceptor fibers by inhibiting voltage-gated ion channels. To further investigate this mechanism
Drug metabolism and disposition: the biological fate of chemicals, 33(12), 1911-1919 (2005-09-27)
In the present study, we investigated the esterase-like activity of human serum albumin (HSA) and the mechanism by which it hydrolyzes, and thereby activates, olmesartan medoxomil (CS-866), a novel angiotensin II receptor antagonist. CS-866 has previously been shown to be
Anesthesia and analgesia, 106(6), 1778-1783 (2008-05-24)
Butamben or n-butyl-p-aminobenzoate is a long-acting experimental local anesthetic for the treatment of chronic pain when given as an epidural suspension. We have investigated whether Cav1.2/L-type calcium channels may be a target of this butamben action. The effect of butamben
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.