Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4688

Sigma-Aldrich

PBPE hydrochloride

≥98% (HPLC)

Synonym(s):

1-[2-[4-(phenylmethyl)phenoxy]ethyl]-Pyrrolidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO·HCl
CAS Number:
Molecular Weight:
317.85
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white

solubility

H2O: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.C1CCN(C1)CCOc2ccc(Cc3ccccc3)cc2

InChI

1S/C19H23NO.ClH/c1-2-6-17(7-3-1)16-18-8-10-19(11-9-18)21-15-14-20-12-4-5-13-20;/h1-3,6-11H,4-5,12-16H2;1H

InChI key

ZWMCGZQOUYHEIQ-UHFFFAOYSA-N

Biochem/physiol Actions

PBPE is a selective ligand of anti-estrogen binding site (AEBS) (Ki=1.2 nM).

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service