Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3937

Sigma-Aldrich

BMX, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ETK, PSCTK2, PSCTK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

29-39 nmol/min·mg

mol wt

~110 kDa

UniProt accession no.

shipped in

dry ice

Storage temp.

−70°C

Gene Information

human ... BMX(660)

Biochem/physiol Actions

The BMX gene encodes a non-receptor tyrosine kinase, which may play a role in the growth and differentiation of hematopoietic cells. The BMX gene is located on chromosomal band Xp22.2 between the DXS197 and DXS207 loci. Interestingly, chromosome X also contains the closest relative of BMX, the BTK gene, implicated in X-linked agammaglobulinemia. BMX is found to induce activation of the Stat signaling pathway.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Tamagnone et al.
Oncogene, 9(12), 3683-3688 (1994-12-01)
The Bmx sequence was identified and cloned during our search for novel tyrosine kinase genes expressed in human bone marrow cells. Bmx cDNA comprises a long open reading frame of 675 amino acids, containing one SH3, one SH2 and one
P Saharinen et al.
Blood, 90(11), 4341-4353 (1997-12-31)
Members of the hematopoietically expressed Tec tyrosine kinase family have an important role in hematopoietic signal transduction, as exemplified by the crucial role of Btk for B-cell differentiation and activation. Although a variety of cell surface receptors have been found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service