Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3935

Sigma-Aldrich

Monoclonal Anti-Bone Morphogenetic Protein 8 antibody produced in mouse

clone 158708, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-BMP-8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

158708, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
western blot: 1-2 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP8B(656)

General description

The anibody shows no cross-reactivity with recombinant human BMP-2, BMP-3, BMP-4, BMP-5, BMP-6, or BMP-7.

Immunogen

purified, E. coli-derived, recombinant mature human bone morphogenetic protein 8.

Biochem/physiol Actions

Bone morphogenetic proteins (BMPs) are the members of the TGF-β superfamily and are identified as the critical factors for formation of bone and cartilage. Apart from this BMPs have immune regulatory functions. The cellular effects of BMPs are mediated by a heteromeric complex composed of type I and type II receptors.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohei Miyazono et al.
Cytokine & growth factor reviews, 16(3), 251-263 (2005-05-06)
Bone morphogenetic proteins (BMPs), members of the transforming growth factor-beta (TGF-beta) superfamily, bind to two different serine/threonine kinase receptors, and mediate their signals through Smad-dependent and Smad-independent pathways. Receptor regulated-Smad (R-Smad) proteins specific for the BMP pathways interact with various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service