B3776
Tamper-evident boxes
Holds 2 tubes or vials up to 17 x 100 mm
All Photos(1)
About This Item
UNSPSC Code:
41121800
Recommended Products
packaging
pkg of 10 ea
size
4 1/4 in. (108 mm) × 1 3/4 in. (44 mm) × 1 in. (25 mm)
General description
For transporting forensic evidence and medical samples with "chain of custody" assurance. Boxes are white plastic, of one-piece construction. They are sealed with plastic ties and numbered locks which must be broken to open.
Packaging
Package contains 10 boxes, 50 locks, and 100 plastic ties.
Choose from one of the most recent versions:
Certificates of Analysis (COA)