Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3760

Sigma-Aldrich

Blue dextran−Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817

form

saline suspension

extent of labeling

1-5 mg per mL

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Application

Blue dextran is used in affinity chromatography as a protein binding site. It has been used to study Alzheimer′s disease by quantifying cerebrospinal fluid (CSF) in relation to amyloid accumulation in rat brain parenchyma.

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Barrett et al.
Infection and immunity, 55(3), 792-802 (1987-03-01)
The presence of proteases in culture supernatant fluids and on the cell surface of Streptococcus sobrinus and the aggregation of multicomponent enzyme complexes make the isolation and characterization of cell surface proteins difficult. We report a simple purification procedure for
W G Deng et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(13), 2463-2470 (2001-11-02)
Salicylic acid (SA), an endogenous signaling molecule of plants, possesses anti-inflammatory and anti-neoplastic actions in human. Its derivative, aspirin, is the most commonly used anti-inflammatory and analgesic drug. Aspirin and sodium salicylate (salicylates) have been reported to have multiple pharmacological
Cerebrospinal fluid Aβ40 and Aβ42: natural course and clinical usefulness.
Shoji, M.
Frontiers in Bioscience, 7, 997-1006 (2002)
R D Law et al.
European journal of biochemistry, 247(2), 642-651 (1997-07-15)
Phosphoenolpyruvate (P-pyruvate) carboxylase from ripened banana fruit was purified to near homogeneity and a final specific activity of 20-23 U/mg protein; P-pyruvate carboxylase-kinase copurified with P-pyruvate carboxylase throughout the purification. Gel filtration FPLC indicated that the two proteins form a
K R Smith et al.
The Journal of biological chemistry, 264(11), 6119-6126 (1989-04-15)
A polypeptide (Mr = 15,000) has been purified from Escherichia coli cell extracts that significantly stimulates the duplex DNA unwinding reaction catalyzed by E. coli Rep protein. The Rep helicase unwinding reaction was stimulated by as much as 20-fold, upon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service