Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3657

Sigma-Aldrich

Restriction Enzyme Buffer SH

10 ×

Synonym(s):

Restriction Endonuclease Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
PubChem Substance ID:

grade

Molecular Biology
for molecular biology

description

10× buffer contains 500 mM Tris-HCl, pH 7.5, 100 mM MgCl2, 1000 mM NaCl, 10 mM dithioerythreitol at 37 °C.

form

buffered aqueous solution

concentration

10 ×

storage temp.

−20°C

SMILES string

CC(C)CC(CNC(Cc1ccccc1)C(N)=O)NC(=O)C(Cc2cnc[nH]2)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(Cc3c[nH]c4ccccc34)NC(=O)C(CCC(N)=O)NC(=O)C(N)Cc5c[nH]c6ccccc56)C(C)C

InChI

1S/C58H77N15O9/c1-32(2)21-39(29-65-46(52(61)76)22-35-13-7-6-8-14-35)69-57(81)48(25-38-28-62-31-67-38)70-50(75)30-66-58(82)51(33(3)4)73-53(77)34(5)68-56(80)47(24-37-27-64-44-18-12-10-16-41(37)44)72-55(79)45(19-20-49(60)74)71-54(78)42(59)23-36-26-63-43-17-11-9-15-40(36)43/h6-18,26-28,31-34,39,42,45-48,51,63-65H,19-25,29-30,59H2,1-5H3,(H2,60,74)(H2,61,76)(H,62,67)(H,66,82)(H,68,80)(H,69,81)(H,70,75)(H,71,78)(H,72,79)(H,73,77)

InChI key

YNZJHLFHFNCZIE-UHFFFAOYSA-N

General description

This buffer is for use with Sigma Restriction Enzymes. Please note every restriction enzyme is supplied with the appropriate buffer - this product is available for users who need additional buffer. For details on buffer composition and protocols for use, see the "Protocols and Articles" tab above.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Roberts
CRC critical reviews in biochemistry, 4(2), 123-164 (1976-11-01)
This review provides a comprehensive account of the current status of the biology and biochemistry of restriction endonucleases. Both Class I and Class II restriction endonucleases will be considered. However, emphasis will be placed on the Class II group, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service