Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3180

Sigma-Aldrich

Apoptosis PCR Primers

Bak primers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

usage

sufficient for 50 PCR reactions

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Synthetic oligonucleotide primers for RT-PCR analysis of mRNA expression. Amplifies reverse-transcript mRNAs from human, mouse and rat sources.

Packaging

Set includes an upstream and a downstream oligonucleotide.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyanka Shankarishan et al.
The Indian journal of medical research, 133, 487-491 (2011-05-31)
Endothelial nitric oxide is a potent vasodilator and impairment of its generation brought about by gene polymorphism is considered a major predictor for several diseases. A single nucleotide polymorphism G894T within exon 7 of endothelial nitric oxide synthase (eNOS-7) gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service