Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2930

Sigma-Aldrich

Bone Morphogenetic Protein Receptor IA/Fc Chimera human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

BMPR-IA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in NSO cells

assay

>90% (SDS-PAGE)

form

lyophilized powder

potency

0.1-0.3 mg per mL ED50

mol wt

subunit calculated mol wt ~41.5 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... BMPR1A(657)

Biochem/physiol Actions

BMPR-IA, also known as activin receptor-like kinase (ALK)-3, is a type I serine/theonine kinase which is required for the signal transduction of the TGF-β family cytokines. Human and murine bone morphogenetic protein receptor IA demonstrates 98% homology.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5.0 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to inhibit recombinant human BMP-4-induced alkaline phosphatase production in C2C12 cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service