Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2685

Sigma-Aldrich

transformation

for protein expression and DNA plasmid production

Synonym(s):

BL21 Competent Cells

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

grade

Molecular Biology

description

T1R

form

suspension

shipped in

dry ice

storage temp.

−70°C

General description

BL21-T1R are competent E. coli that are suitable for high level expression/production of heterologous proteins regulated by various expression vector systems. The cells have transformation efficiency of ≥3x106 cfu/μg when transformed with non-saturating amounts of pUC19 plasmid DNA.
Sigma′s BL21-T1R competent E. coli cells are grown and made chemically competent using an optimized procedure specific to the strain, followed by strain verification and efficiency testing. The cells are provided in frozen 50 μl aliquots for convenience. Each aliquot can be used for a single transformation.

Application

Suitable for expression and production of proteins directed by a range of expression systems with promoters such as lac, trc, tac, λPL and araD

Features and Benefits

  • Ensures recovery of heterologous proteins
  • Contains the genotype tonA that protects the clonal stocks from lytic bacteriophages
  • Guaranteed high transformation efficiency
  • Convenient 50 μL aliquots

Components

  • BL21-T1R chemically competent cells, 10 X 50 μL (B2810)
  • pUC 19 control DNA (10 ng/μL), 10 μL (D2567)

Principle

BL21-T1R does not express ion proteases and outer membrane protease, ompT. This prevents the degradation of heterologous proteins expressed by various expression vector systems. This strain also contains tonA genotype that confers resistance to lytic bacteriophages such as T1 and T5.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

new publication: Bacterial receptors for phages and colicins as constituents of specific transport system, in Receptors and Recognition, Series B, Vol. 3, J. L. Reissig (Ed.), pp. 101-130. (Chapman & Hall, Ltd., London, 1977).
Braun, V., and Hantke, K.
Receptors and Recognition, 3, 101-130 (1977)
Host specificity of DNA produced by Escherichia coli: bacterial mutations affecting the restriction and modification of DNA.
W B Wood
Journal of molecular biology, 16(1), 118-133 (1966-03-01)
T A Phillips et al.
Journal of bacteriology, 159(1), 283-287 (1984-07-01)
The product of the pleiotropic gene lon is a protein with protease activity and has been tentatively identified as protein H94.0 on the reference two-dimensional gel of Escherichia coli proteins. Purified Lon protease migrated with the prominent cellular protein H94.0
Losee L Ling et al.
Nature, 517(7535), 455-459 (2015-01-07)
Antibiotic resistance is spreading faster than the introduction of new compounds into clinical practice, causing a public health crisis. Most antibiotics were produced by screening soil microorganisms, but this limited resource of cultivable bacteria was overmined by the 1960s. Synthetic
Adam Johnson et al.
The EMBO journal, 34(6), 811-827 (2015-01-15)
In mammalian cells, cargo-laden secretory vesicles leave the endoplasmic reticulum (ER) en route to ER-Golgi intermediate compartments (ERGIC) in a manner dependent on the COPII coat complex. We report here that COPII-coated transport carriers traverse a submicron, TFG (Trk-fused gene)-enriched

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service