Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2519

Sigma-Aldrich

O-t-Butyl-L-tyrosine t-butyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO3 · HCl
CAS Number:
Molecular Weight:
329.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

storage temp.

−20°C

SMILES string

Cl.CC(C)(C)OC(=O)[C@@H](N)Cc1ccc(OC(C)(C)C)cc1

InChI

1S/C17H27NO3.ClH/c1-16(2,3)20-13-9-7-12(8-10-13)11-14(18)15(19)21-17(4,5)6;/h7-10,14H,11,18H2,1-6H3;1H/t14-;/m0./s1

InChI key

ZAHGOULTAJWDGV-UQKRIMTDSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Cassano et al.
International journal of pharmaceutics, 589, 119829-119829 (2020-09-03)
This work aims to the synthesis of novel carboxylated chitosan-dopamine (DA) and -tyrosine (Tyr) conjugates as systems for improving the brain delivery of the neurotransmitter DA following nasal administration. For this purpose, ester or amide conjugates were synthesized by N,N-dicyclohexylcarbodiimide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service