Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B1779

Sigma-Aldrich

Borage oil from Borago officinalis seeds

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211

form

liquid

lipid type

oils

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oil derived from the seeds of Borago officinalis. Highest known plant source of γ-linolenic acid. Anti-inflammatory. Used as an alternative treatment for gastrointestinal, respiratory, and cardiovascular disorders.

Other Notes

Contains 20-25% γ-linolenic acid (as triglycerides)

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service