Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B1426

Millipore

Bismuth sulphite agar

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

powder

pH

7.7±0.2 (25 °C)

application(s)

microbiology

Application

Recommended for the selective isolation and preliminary identification of Salmonella typhi and other Salmonellae.

Other Notes

Components (g/L)
Peptic Digest of Animal Tissue 10.00
Beef Extract 5.00
Dextrose 5.00
Disodium Phosphate 4.00
Ferrous Sulphate 0.30
Bismuth Sulphite Indicator 8.00
Brilliant Green 0.025
Agar 20.00

replaced by

hcodes

Hazard Classifications

Aquatic Chronic 3

supp_hazards

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Washington J.A.
Laboratory Procedures in Clinical Microbiology (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service