Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B0809

Sigma-Aldrich

Monoclonal Anti-Bcl-w antibody produced in mouse

clone 90316.11, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

90316.11, monoclonal

form

buffered aqueous solution

mol wt

antigen 18 kDa by SDS-PAGE

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL using human WS1cells or mouse CTLL2 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL2L2(599)
mouse ... Bcl2l2(12050)

Immunogen

purified recombinant human Bcl-w amino acids 1-172 with 6 histidines at the C‑terminus.

Physical form

Supplied as a 0.2 μm filtered solution in phosphate buffered saline and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service