Key Documents
B0783
Nalgene® centrifuge bottles, Style 3122
nominal capacity 1000 mL, for IEC rotors
Synonym(s):
nalgene bottle, polycarbonate bottle
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
polycarbonate
round bottle
screw top cap
translucent bottle
translucent polycarbonate
translucent polypropylene screw closure
joint
Joints threaded neck (52.6)
feature
RCF 7,100 × g
with cap
packaging
pack of 4
manufacturer/tradename
Nalgene® 3122-1010
parameter
7100 × g max. RCF
bottle diam. × L
69.52 mm × 158.9 mm
nominal capacity
1000 mL
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)