Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B0777

Sigma-Aldrich

Brain natriuretic peptide fragment 7-32

≥97% (HPLC)

Synonym(s):

BNP-26

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C120H198N42O36S2
CAS Number:
Molecular Weight:
2869.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97% (HPLC)

form

solid

composition

Peptide content, ~65%

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CO)NC(=O)CNC1=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc3ccc(O)cc3)C(O)=O)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@@H](N)CC(O)=O

InChI

1S/C120H198N42O36S2/c1-13-62(12)94-113(195)142-51-88(170)145-82(54-165)110(192)153-74(40-59(6)7)107(189)160-81(53-164)99(181)140-48-87(169)144-72(38-57(2)3)96(178)138-49-89(171)147-84(112(194)155-77(45-85(122)167)109(191)161-93(61(10)11)114(196)157-75(41-60(8)9)105(187)150-68(25-18-34-134-117(125)126)101(183)149-70(27-20-36-136-119(129)130)103(185)158-79(115(197)198)43-64-29-31-65(166)32-30-64)56-200-199-55-83(146-90(172)50-141-98(180)80(52-163)159-95(177)66(121)44-91(173)174)111(193)154-76(42-63-22-15-14-16-23-63)97(179)139-47-86(168)143-67(24-17-33-133-116(123)124)100(182)148-69(26-19-35-135-118(127)128)102(184)152-73(39-58(4)5)106(188)156-78(46-92(175)176)108(190)151-71(104(186)162-94)28-21-37-137-120(131)132/h14-16,22-23,29-32,57-62,66-84,93-94,163-166H,13,17-21,24-28,33-56,121H2,1-12H3,(H2,122,167)(H,138,178)(H,139,179)(H,140,181)(H,141,180)(H,142,195)(H,143,168)(H,144,169)(H,145,170)(H,146,172)(H,147,171)(H,148,182)(H,149,183)(H,150,187)(H,151,190)(H,152,184)(H,153,192)(H,154,193)(H,155,194)(H,156,188)(H,157,196)(H,158,185)(H,159,177)(H,160,189)(H,161,191)(H,162,186)(H,173,174)(H,175,176)(H,197,198)(H4,123,124,133)(H4,125,126,134)(H4,127,128,135)(H4,129,130,136)(H4,131,132,137)/t62-,66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,93-,94-/m0/s1

InChI key

XDUBCYDPVVBFSE-CVWICPBCSA-N

Application

Useful as an immunogen for preparing anti-BNP antibodies

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Sudoh et al.
Nature, 332(6159), 78-81 (1988-03-03)
Atrial natriuretic peptide (ANP), a hormone secreted from mammalian atria, regulates the homoeostatic balance of body fluid and blood pressure. ANP-like immunoreactivity is also present in the brain, suggesting that the peptide functions as a neuropeptide. We report here identification
Abdu Adem et al.
PloS one, 8(3), e57806-e57806 (2013-03-22)
The objectives of this study were to investigate and compare the responses of atrial natriuretic peptide (ANP) and B-type natriuretic peptide (BNP) in the circulation of hydrated, dehydrated, and dehydrated losartan - treated camels; and to document the cardiac storage
Yanhao Zhang et al.
Reproductive biology and endocrinology : RB&E, 15(1), 41-41 (2017-06-01)
Natriuretic peptides (NPs), brain and C type NPs (BNP and CNP), were involved in the maintenance of porcine oocyte meiotic arrest. The present study investigated the effects of NPs on developmental competence of immature porcine oocytes with follicles of different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service