Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B0686

Sigma-Aldrich

Anti-BTK, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Agammaglobulinemia tyrosine kinase (ATK), Anti-Bruton’s tyrosine kinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 75 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

microarray: suitable
western blot: 1-2 μg/mL using lysates of Ramos (Human Burkitt lymphoma) cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BTK(695)

General description

BTK is a tyrosine-protein kinase that modulates signaling, growth, and differentiation in B-lymphocytes. BTK also mediates cytoskeletal rearrangements and associates with Pin1 to regulate the cell-cycle dependent expression of BTK. Additionally, BTK signaling cascades are involved in regulation of calcium fluxes and transcription. Genetic alterations in BTK1 have been linked to X-linked aγglobulinemia . Anti-BTK, N-Terminal antibody is specific for human BTK (75 kDa). In immunoblotting, staining of the BTK band is specifically inhibited by the immunizing peptide.

Immunogen

The immunogen sequence is conserved in mouse, rat and chicken.
synthetic peptide corresponding to amino acids 11-26 of human BTK, conjugated to KLH via a C-terminal added cysteine residue. This sequence differs from the human Tec tyrosine kinase by 2 amino acids.

Application

Anti-BTK, N-Terminal antibody is suitable for use in western blot (1-2 μg/mL using lysates of Ramos (Human Burkitt lymphoma) cell line) and microarray.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B F Nore et al.
European journal of immunology, 30(1), 145-154 (1999-12-22)
Bruton's tyrosine kinase (Btk) is a member of the Tec family of protein tyrosine kinases (PTK) characterized by an N-terminal pleckstrin homology domain (PH) thought to directly interact with phosphoinositides. We report here that wild-type (wt) and also a gain-of-function
Liang Yu et al.
The Journal of biological chemistry, 281(26), 18201-18207 (2006-04-29)
Bruton tyrosine kinase (Btk) is expressed in B-lymphocytes. Mutations in Btk cause X-linked agammaglobulinemia in humans. However, the mechanism of activation and signaling of this enzyme has not been fully investigated. We have here shown that the peptidylprolyl cis/trans isomerase
Abdalla J Mohamed et al.
Immunological reviews, 228(1), 58-73 (2009-03-18)
Bruton's agammaglobulinemia tyrosine kinase (Btk) is a cytoplasmic tyrosine kinase important in B-lymphocyte development, differentiation, and signaling. Btk is a member of the Tec family of kinases. Mutations in the Btk gene lead to X-linked agammaglobulinemia (XLA) in humans and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service