Key Documents
B0305
Sigma-Aldrich
Ohaus® Compact portable balance
Ohaus® CS 2000
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
material
stainless steel weighing plate
feature
CE compliant
CSA approved
manufacturer/tradename
Ohaus® CS 2000
weighing capacity measuring range
2000 g, precision: 1.0 g
diam. × diam.
145 x 133 mm × 5.75 x 5.25 in.
battery power
3 AA (alkaline batteries (included))
General description
Power: 3-AA alkaline batteries (included) or AC adapter (optional); Display: LCD, Units: pounds, grams, ounces; platform: 5.25 × 4.75 in., dimensions: 7.6 × 5.3 × 1.5 in.
Features: Low battery indicator, overload and underload indicators, auto shut-off, rugged plastic construction, tactile feedback keypad.
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)