AXYZT50LR
MAXYMum Recovery® pipet filter tips for Zymark® automated liquid handling system
size 50 μL, pkg of 5x960tips/cs, clear, racked
Synonym(s):
pipette tip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.02
volume:
50 μL
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc ZT-50-L-R
Corning
Corning
manufacturer/tradename
Axygen Inc ZT-50-L-R
Corning
packaging
pkg of 5x960tips/cs
size
50 μL
volume
50 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Legal Information
Maxymum Recovery is a registered trademark of Corning, Inc.
Zymark is a registered trademark of Sotax Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service