Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYTTF1000CBKHTRS

Tecan®-style liquid level sensing filter tips

size 1000 μL, case of 1538 ea 16x96tips/cs, racked, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$164.79

$164.79

List Price$289.00Save 43%
Web-Only Promotion

Check Cart for Availability


Select a Size

Change View
1 PKG
$164.79

About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
1000 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier
endotoxin free
filter
packaging:
case of 1538 ea 16x96tips/cs
manufacturer/tradename:
Corning TTF-1000-CBK-HTR-S

$164.79

List Price$289.00Save 43%
Web-Only Promotion

Check Cart for Availability

material

black

sterility

sterile

feature

DNA free
RNase free
barrier
endotoxin free
filter

packaging

case of 1538 ea 16x96tips/cs

manufacturer/tradename

Corning TTF-1000-CBK-HTR-S

size

1000 μL

volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

Legal Information

Tecan is a registered trademark of Tecan Group AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service